siRNA Details
| Gene Symbol: Ngb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033359.3 | NP_203523.2 | 2, 3 | 618 | ||
| XM_008764864.1 | XP_008763086.1 | 2, 3 | 1299 | |||
| GenBank mRNA | AB056655.1 | - | 2, 3 | 190 | ||
| AF333245.1 | - | 2, 3 | 209 | |||
| AF334379.1 | - | 2, 3 | 209 | |||
| AY066001.1 | - | 2, 3 | 418 | |||

Ngb
Genomic Map