siRNA Details
| Gene Symbol: Plk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031821.1 | NP_114009.1 | 3, 4 | 624 | ||
| GenBank mRNA | AF136583.1 | - | 3, 4 | 624 | ||
| BC070878.1 | - | 3, 4 | 608 | 7096750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plk2| Gene Symbol: Plk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031821.1 | NP_114009.1 | 3, 4 | 624 | ||
| GenBank mRNA | AF136583.1 | - | 3, 4 | 624 | ||
| BC070878.1 | - | 3, 4 | 608 | 7096750 | ||