siRNA Details
| Gene Symbol: Cyp2c23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031839.3 | NP_114027.2 | 2, 3 | 368 | ||
| GenBank mRNA | BC078707.1 | - | 2, 3 | 368 | 7110580 | |
| U04733.1 | - | 2, 3 | 349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2c23| Gene Symbol: Cyp2c23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031839.3 | NP_114027.2 | 2, 3 | 368 | ||
| GenBank mRNA | BC078707.1 | - | 2, 3 | 368 | 7110580 | |
| U04733.1 | - | 2, 3 | 349 | |||