siRNA Details
| Gene Symbol: Fdps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031840.1 | NP_114028.1 | 2 | 279 | ||
| GenBank mRNA | BC059125.1 | - | 2 | 207 | 6919326 | |
| FQ209697.1 | - | 2 | 172 | |||
| FQ210400.1 | - | 2 | 171 | |||
| FQ211073.1 | - | 3 | 388 | |||
| FQ213475.1 | - | 2 | 203 | |||
| FQ218913.1 | - | 2 | 173 | |||
| FQ218933.1 | - | 2 | 173 | |||
| FQ219092.1 | - | 2 | 173 | |||
| FQ225110.1 | - | 2 | 241 | |||
| M17300.1 | - | 1 | 149 | |||
| M34477.1 | - | 2 | 279 | |||

Fdps
Genomic Map