siRNA Details
| Gene Symbol: Prkab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031976.1 | NP_114182.1 | 2 | 402 | ||
| XM_006249481.2 | XP_006249543.1 | 3 | 402 | |||
| GenBank mRNA | BC062008.1 | - | 2 | 288 | 5599996 | |
| U42411.1 | - | 2 | 402 | |||
| X95577.1 | - | 3 | 372 | |||

Prkab1
Genomic Map