siRNA Details
| Gene Symbol: Crot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031987.1 | NP_114193.1 | 3, 4 | 285 | ||
| XM_006236004.2 | XP_006236066.1 | 3, 4 | 445 | |||
| XM_006236005.2 | XP_006236067.1 | 4, 5 | 1775 | |||
| GenBank mRNA | BC074004.1 | - | 3, 4 | 305 | 7098081 | |
| J02844.1 | - | 726 | ||||
| U26033.1 | - | 3, 4 | 285 | |||

Crot
Genomic Map