siRNA Details
| Gene Symbol: BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005180.8 | NP_005171.4 | 3, 4 | 723 | ||
| GenBank mRNA | BC011652.2 | - | 3, 4 | 722 | 4138748 | |
| BT019450.1 | - | 2, 3 | 217 | |||
| CR456911.1 | - | 2, 3 | 217 | |||
| L13689.1 | - | 3, 4 | 696 | |||
| Gene Symbol: COMMD3-BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204062.1 | NP_001190991.1 | 7, 8 | 681 | ||
| GenBank mRNA | AK313235.1 | - | 7, 8 | 656 | ||

BMI1
Genomic Map