siRNA Details
| Gene Symbol: Hsd17b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032066.1 | NP_114455.1 | 3 | 269 | ||
| XM_006234599.2 | XP_006234661.1 | 3 | 436 | |||
| GenBank mRNA | BC061543.1 | - | 3 | 290 | 6888949 | |
| FQ213557.1 | - | 3 | 305 | |||
| FQ214777.1 | - | 3 | 334 | |||
| U81186.1 | - | 3 | 269 | |||

Hsd17b12
Genomic Map