siRNA Details
Gene Symbol: Irs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032074.1 | NP_114463.1 | 2 | 1153 | ||
GenBank mRNA | BC161840.1 | - | 2 | 1005 | 9027966 | |
U93880.1 | - | 2 | 1153 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032074.1 | NP_114463.1 | 2 | 1153 | ||
GenBank mRNA | BC161840.1 | - | 2 | 1005 | 9027966 | |
U93880.1 | - | 2 | 1153 |