siRNA Details
| Gene Symbol: Pld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033299.2 | NP_150641.2 | 2, 3 | 437 | ||
| XM_008767778.1 | XP_008766000.1 | 2, 3 | 479 | |||
| GenBank mRNA | AB003172.1 | - | 3, 4 | 401 | ||
| D88672.1 | - | 2, 3 | 437 | |||

Pld2
Genomic Map