siRNA Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031820.1 | NP_114008.1 | 8, 9 | 918 | ||
| XM_006239598.2 | XP_006239660.1 | 8, 9 | 1232 | |||
| XM_008764388.1 | XP_008762610.1 | 7, 8 | 1320 | |||
| XM_008764389.1 | XP_008762611.1 | 7, 8 | 1320 | |||
| XM_008764390.1 | XP_008762612.1 | 7, 8 | 1320 | |||
| XM_008764391.1 | XP_008762613.1 | 8, 9 | 1021 | |||
| GenBank mRNA | AF143545.2 | - | 8, 9 | 918 | ||
| AF143546.2 | - | 8, 9 | 918 | |||
| BC160920.1 | - | 1, 2 | 67 | 7461890 | ||

Dvl1
Genomic Map