siRNA Details
| Gene Symbol: Fbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031825.1 | NP_114013.1 | 17, 18 | 2172 | ||
| XM_006234935.2 | XP_006234997.1 | 18, 19 | 2461 | |||
| XM_008762202.1 | XP_008760424.1 | 17, 18 | 2362 | |||
| XM_008762203.1 | XP_008760425.1 | 18, 19 | 2456 | |||
| GenBank mRNA | AF135059.1 | - | 17, 18 | 2172 | ||

Fbn1
Genomic Map