siRNA Details
| Gene Symbol: Src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031977.1 | NP_114183.1 | 6 | 620 | ||
| XM_008762387.1 | XP_008760609.1 | 6 | 629 | |||
| XM_008762388.1 | XP_008760610.1 | 6 | 666 | |||
| XM_008762389.1 | XP_008760611.1 | 6 | 737 | |||
| GenBank mRNA | AF130457.1 | - | 5 | 602 | ||
| AF157016.1 | - | 6 | 620 | |||
| DQ120509.1 | - | 5 | 586 | |||
| DQ120510.1 | - | 5 | 586 | |||

Src
Genomic Map