siRNA Details
| Gene Symbol: Ybx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031979.3 | NP_114185.3 | 8 | 1079 | ||
| GenBank mRNA | D28557.1 | - | 7 | 736 | ||
| FQ226232.1 | - | 5 | 240 | |||
| U22893.1 | - | 8 | 1082 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ybx3| Gene Symbol: Ybx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031979.3 | NP_114185.3 | 8 | 1079 | ||
| GenBank mRNA | D28557.1 | - | 7 | 736 | ||
| FQ226232.1 | - | 5 | 240 | |||
| U22893.1 | - | 8 | 1082 | |||