siRNA Details
| Gene Symbol: Sdcbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031986.1 | NP_114192.1 | 5 | 417 | ||
| GenBank mRNA | AF248548.1 | - | 5 | 417 | ||
| AJ292243.1 | - | 4 | 360 | |||
| BC064651.1 | - | 5 | 385 | 5598152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdcbp| Gene Symbol: Sdcbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031986.1 | NP_114192.1 | 5 | 417 | ||
| GenBank mRNA | AF248548.1 | - | 5 | 417 | ||
| AJ292243.1 | - | 4 | 360 | |||
| BC064651.1 | - | 5 | 385 | 5598152 | ||