siRNA Details
| Gene Symbol: Crot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031987.1 | NP_114193.1 | 7, 8 | 809 | ||
| XM_006236004.2 | XP_006236066.1 | 7, 8 | 969 | |||
| XM_006236005.2 | XP_006236067.1 | 8, 9 | 2299 | |||
| GenBank mRNA | BC074004.1 | - | 7, 8 | 829 | 7098081 | |
| J02844.1 | - | 1250 | ||||
| U26033.1 | - | 7, 8 | 809 | |||

Crot
Genomic Map