siRNA Details
| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032062.2 | NP_114451.2 | 27, 28 | 4259 | ||
| GenBank mRNA | AF229255.1 | - | 18, 19 | 2920 | ||
| AF230644.1 | - | 27, 28 | 4259 | |||
| AF232668.1 | - | 26, 27 | 4229 | |||
| AF232669.1 | - | 26, 27 | 4229 | |||
| U88156.1 | - | 4612 | ||||
| U88157.1 | - | 26, 27 | 4229 | |||

Kalrn
Genomic Map