siRNA Details
Gene Symbol: Nae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127481.1 | - | 10 | 774 | 8370668 | |
FM131178.1 | - | 11 | ||||
U90829.1 | - | 10 | 771 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127481.1 | - | 10 | 774 | 8370668 | |
FM131178.1 | - | 11 | ||||
U90829.1 | - | 10 | 771 |