siRNA Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052798.2 | NP_434685.1 | 2, 3 | 480 | ||
| XM_006246227.2 | XP_006246289.1 | 3, 4 | 1627 | |||
| GenBank mRNA | M96548.1 | - | 2, 3 | 482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp354a| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052798.2 | NP_434685.1 | 2, 3 | 480 | ||
| XM_006246227.2 | XP_006246289.1 | 3, 4 | 1627 | |||
| GenBank mRNA | M96548.1 | - | 2, 3 | 482 | ||