siRNA Details
| Gene Symbol: Chrna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052805.2 | NP_434692.2 | 2, 3 | 209 | ||
| GenBank mRNA | AY574254.1 | - | 2, 3 | 209 | ||
| L31621.1 | - | 2, 3 | 285 | |||
| X03440.1 | - | 2, 3 | 359 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145129, BC120599, AK053497, AK080475, AK051730, and others.

Chrna3
Genomic Map