siRNA Details
| Gene Symbol: Bpifa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052808.1 | NP_434695.1 | 4 | 378 | ||
| XM_008762379.1 | XP_008760601.1 | 5 | 727 | |||
| GenBank mRNA | M83209.1 | - | 4 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifa2| Gene Symbol: Bpifa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052808.1 | NP_434695.1 | 4 | 378 | ||
| XM_008762379.1 | XP_008760601.1 | 5 | 727 | |||
| GenBank mRNA | M83209.1 | - | 4 | 378 | ||