siRNA Details
| Gene Symbol: Hfe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053301.4 | NP_445753.1 | 2 | 247 | ||
| GenBank mRNA | AF008587.1 | - | 1 | 46 | ||
| AJ001517.1 | - | 2 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hfe| Gene Symbol: Hfe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053301.4 | NP_445753.1 | 2 | 247 | ||
| GenBank mRNA | AF008587.1 | - | 1 | 46 | ||
| AJ001517.1 | - | 2 | 286 | |||