siRNA Details
| Gene Symbol: Ip6k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053316.2 | NP_445768.1 | 2, 3 | 385 | ||
| XM_006243839.1 | XP_006243901.1 | 2, 3 | 587 | |||
| GenBank mRNA | AB049151.1 | - | 2, 3 | 436 | ||
| BC078702.1 | - | 2, 3 | 385 | 7132706 | ||
| FQ211545.1 | - | 2, 3 | 533 | |||

Ip6k1
Genomic Map