siRNA Details
| Gene Symbol: Nr0b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053317.1 | NP_445769.1 | 1 | 443 | ||
| GenBank mRNA | X99470.1 | - | 1 | 443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr0b1| Gene Symbol: Nr0b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053317.1 | NP_445769.1 | 1 | 443 | ||
| GenBank mRNA | X99470.1 | - | 1 | 443 | ||