siRNA Details
| Gene Symbol: Syt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053324.1 | NP_445776.1 | 3 | 575 | ||
| XM_006229961.2 | XP_006230023.1 | 3 | 626 | |||
| XM_008759790.1 | XP_008758012.1 | 3 | 626 | |||
| GenBank mRNA | AF375461.1 | - | 3 | 575 | ||

Syt9
Genomic Map