siRNA Details
| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053339.1 | NP_445791.1 | 2 | 147 | ||
| XM_008770372.1 | XP_008768594.1 | 2 | 2853 | |||
| XM_008770373.1 | XP_008768595.1 | 2 | 2896 | |||
| XM_008770374.1 | XP_008768596.1 | 1 | 120 | |||
| GenBank mRNA | X95188.1 | - | 2 | 147 | ||

Acox3
Genomic Map