siRNA Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053343.3 | NP_445795.1 | 4 | 368 | ||
| XM_008761004.1 | XP_008759226.1 | 4 | 2270 | |||
| XM_008761005.1 | XP_008759227.1 | 4 | 1296 | |||
| XM_008761006.1 | XP_008759228.1 | 4 | 2273 | |||
| XM_008761007.1 | XP_008759229.1 | 4 | 380 | |||
| GenBank mRNA | FQ211799.1 | - | 4 | 355 | ||
| U78857.1 | - | 4 | 358 | |||

Dclk1
Genomic Map