siRNA Details
| Gene Symbol: Ctnnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053357.2 | NP_445809.2 | 2 | 161 | ||
| XM_008766691.1 | XP_008764913.1 | 2 | 244 | |||
| GenBank mRNA | AF121265.1 | - | 2 | 161 | ||
| FQ222424.1 | - | 354 | ||||
| FQ224468.1 | - | 359 | ||||

Ctnnb1
Genomic Map