siRNA Details
| Gene Symbol: Hps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_040669.1 | NP_414541.1 | 5 | 506 | ||
| XM_006231378.2 | XP_006231440.1 | 6 | 633 | |||
| XM_006231379.2 | XP_006231441.1 | 6 | 759 | |||
| XM_006231380.2 | XP_006231442.1 | 6 | 589 | |||
| XM_006231381.2 | XP_006231443.1 | 6 | 700 | |||
| GenBank mRNA | AF333325.1 | - | 5 | 506 | ||

Hps1
Genomic Map