siRNA Details
| Gene Symbol: Cxcr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053303.1 | NP_445755.1 | 2 | 581 | ||
| XM_006242901.2 | XP_006242963.1 | 3 | 1204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcr5| Gene Symbol: Cxcr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053303.1 | NP_445755.1 | 2 | 581 | ||
| XM_006242901.2 | XP_006242963.1 | 3 | 1204 | |||