siRNA Details
| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053307.2 | NP_445759.1 | 3 | 271 | ||
| XM_008770745.1 | XP_008768967.1 | 3 | 642 | |||
| GenBank mRNA | AY005464.1 | - | 3 | 261 | ||
| BC087009.1 | - | 3 | 271 | 7107102 | ||
| DQ989019.1 | - | 4 | 303 | |||
| DQ989020.1 | - | 4 | 306 | |||
| DQ989021.1 | - | 3 | 237 | |||

Msra
Genomic Map