siRNA Details
| Gene Symbol: Ager | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053336.2 | NP_445788.2 | 3 | 231 | ||
| XM_006256025.1 | XP_006256087.1 | 3 | 214 | |||
| GenBank mRNA | BC084697.1 | - | 3 | 213 | 7121345 | |
| GU164715.1 | - | 2 | 398 | |||
| GU164716.1 | - | 2 | 398 | |||
| GU164717.1 | - | 3 | 197 | |||
| GU164718.1 | - | 2 | 398 | |||
| GU164719.1 | - | 3 | 196 | |||
| GU164725.1 | - | 3 | 197 | |||
| L33413.1 | - | 3 | 231 | |||

Ager
Genomic Map