siRNA Details
| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053373.1 | NP_445825.1 | 2 | 294 | ||
| GenBank mRNA | AF154114.1 | - | 2 | 294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pglyrp1| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053373.1 | NP_445825.1 | 2 | 294 | ||
| GenBank mRNA | AF154114.1 | - | 2 | 294 | ||