siRNA Details
| Gene Symbol: Gjb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053388.1 | NP_445840.1 | 3 | 609 | ||
| XM_006252079.2 | XP_006252141.1 | 3 | 1935 | |||
| XM_006252080.2 | XP_006252142.1 | 3 | 1511 | |||
| XM_008770781.1 | XP_008769003.1 | 3 | 723 | |||
| XM_008770782.1 | XP_008769004.1 | 3 | 1033 | |||
| XM_008770783.1 | XP_008769005.1 | 3 | 614 | |||
| XM_008770784.1 | XP_008769006.1 | 3 | 720 | |||
| GenBank mRNA | AF170284.1 | - | 3 | 609 | ||
| BC078753.1 | - | 3 | 606 | 7132296 | ||

Gjb6
Genomic Map