siRNA Details
| Gene Symbol: Nos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052799.1 | NP_434686.1 | 1 | 412 | ||
| XM_008769210.1 | XP_008767432.1 | 2 | 1223 | |||
| GenBank mRNA | U67309.1 | - | 1 | 412 | ||
| X59949.1 | - | 1 | 659 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nos1| Gene Symbol: Nos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052799.1 | NP_434686.1 | 1 | 412 | ||
| XM_008769210.1 | XP_008767432.1 | 2 | 1223 | |||
| GenBank mRNA | U67309.1 | - | 1 | 412 | ||
| X59949.1 | - | 1 | 659 | |||