siRNA Details
| Gene Symbol: Cdo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052809.1 | NP_434696.1 | 3 | 495 | ||
| XM_006254698.2 | XP_006254760.1 | 3 | 720 | |||
| GenBank mRNA | BC070509.1 | - | 3 | 486 | 7100321 | |
| FQ218376.1 | - | 2 | 132 | |||
| FQ219282.1 | - | 2 | 132 | |||
| M35266.1 | - | 3 | 495 | |||

Cdo1
Genomic Map