siRNA Details
| Gene Symbol: Slc5a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052983.2 | NP_443215.2 | 9, 10 | 1257 | ||
| GenBank mRNA | U60282.1 | - | 9, 10 | 1282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a5| Gene Symbol: Slc5a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052983.2 | NP_443215.2 | 9, 10 | 1257 | ||
| GenBank mRNA | U60282.1 | - | 9, 10 | 1282 | ||