siRNA Details
| Gene Symbol: Ubd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053299.2 | NP_445751.2 | 2 | 404 | ||
| GenBank mRNA | AJ312394.1 | - | 2 | 406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubd| Gene Symbol: Ubd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053299.2 | NP_445751.2 | 2 | 404 | ||
| GenBank mRNA | AJ312394.1 | - | 2 | 406 | ||