siRNA Details
| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053307.2 | NP_445759.1 | 5, 6 | 555 | ||
| GenBank mRNA | AY005464.1 | - | 5, 6 | 545 | ||
| BC087009.1 | - | 5, 6 | 555 | 7107102 | ||
| DQ989019.1 | - | 6, 7 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msra| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053307.2 | NP_445759.1 | 5, 6 | 555 | ||
| GenBank mRNA | AY005464.1 | - | 5, 6 | 545 | ||
| BC087009.1 | - | 5, 6 | 555 | 7107102 | ||
| DQ989019.1 | - | 6, 7 | 587 | |||