siRNA Details
| Gene Symbol: Hpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053318.1 | NP_445770.1 | 6, 7 | 730 | ||
| GenBank mRNA | BC091137.1 | - | 6, 7 | 721 | 7326169 | |
| FQ210231.1 | - | 6, 7 | 731 | |||
| FQ211176.1 | - | 6, 7 | 732 | |||
| FQ218971.1 | - | 6, 7 | 723 | |||
| FQ218998.1 | - | 3, 4 | 499 | |||
| FQ219255.1 | - | 1, 2 | 179 | |||
| FQ219309.1 | - | 6, 7 | 729 | |||

Hpx
Genomic Map