siRNA Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053327.1 | NP_445779.1 | 16 | 1746 | ||
| XM_008764291.1 | XP_008762513.1 | 16 | 1862 | |||
| XM_008764292.1 | XP_008762514.1 | 16 | 1832 | |||
| GenBank mRNA | BC081761.1 | - | 16 | 1812 | 7108273 | |
| D13927.1 | - | 16 | 1746 | |||
| Z34291.1 | - | 15 | 1703 | |||

Clcnka
Genomic Map