siRNA Details
| Gene Symbol: MTMR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004685.3 | NP_004676.3 | 6 | 975 | ||
| XM_011535306.1 | XP_011533608.1 | 6 | 921 | |||
| XM_011535307.1 | XP_011533609.1 | 6 | 921 | |||
| XM_017020845.1 | XP_016876334.1 | 6 | 921 | |||
| XM_017020846.1 | XP_016876335.1 | 2 | 126 | |||
| GenBank mRNA | AF072928.1 | - | 2 | 189 | ||
| AF406619.1 | - | 6 | 656 | |||
| AK314587.1 | - | 6 | 840 | |||
| AL832017.2 | - | 6 | 898 | |||
| BC040012.1 | - | 6 | 847 | 5495564 | ||
| EF491793.1 | - | 1 | 45 | |||

MTMR6
Genomic Map