siRNA Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053343.3 | NP_445795.1 | 10, 11 | 1096 | ||
| XM_008761004.1 | XP_008759226.1 | 10, 11 | 2998 | |||
| XM_008761005.1 | XP_008759227.1 | 10, 11 | 2024 | |||
| XM_008761006.1 | XP_008759228.1 | 10, 11 | 3001 | |||
| XM_008761007.1 | XP_008759229.1 | 10, 11 | 1108 | |||
| GenBank mRNA | CB586027.1 | - | 3, 4 | 274 | ||
| FQ211799.1 | - | 10, 11 | 1083 | |||
| U78857.1 | - | 10, 11 | 1086 | |||

Dclk1
Genomic Map