siRNA Details
| Gene Symbol: Slpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053372.1 | NP_445824.1 | 3 | 300 | ||
| GenBank mRNA | AF151982.1 | - | 3 | 300 | ||
| AF178426.1 | - | 3 | 299 | |||
| AF421377.1 | - | 3 | 310 | |||
| Gene Symbol: LOC103690289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762552.1 | XP_008760774.1 | 3 | 291 | ||
| Gene Symbol: RGD1563818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235625.2 | XP_006235687.1 | 3 | 291 | ||

Slpi
Genomic Map