siRNA Details
| Gene Symbol: Atp1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053381.1 | NP_445833.1 | 7 | 905 | ||
| GenBank mRNA | AF158385.2 | - | 7 | 905 | ||
| AF158386.2 | - | 7 | 893 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp1b4| Gene Symbol: Atp1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053381.1 | NP_445833.1 | 7 | 905 | ||
| GenBank mRNA | AF158385.2 | - | 7 | 905 | ||
| AF158386.2 | - | 7 | 893 | |||