siRNA Details
| Gene Symbol: Gemin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053389.1 | NP_445841.1 | 5 | 481 | ||
| GenBank mRNA | AF176072.1 | - | 5 | 481 | ||
| BC098662.1 | - | 5 | 424 | 7386123 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gemin2| Gene Symbol: Gemin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053389.1 | NP_445841.1 | 5 | 481 | ||
| GenBank mRNA | AF176072.1 | - | 5 | 481 | ||
| BC098662.1 | - | 5 | 424 | 7386123 | ||