siRNA Details
| Gene Symbol: Hs3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053391.1 | NP_445843.1 | 1 | 119 | ||
| GenBank mRNA | AF177430.1 | - | 1 | 119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st1| Gene Symbol: Hs3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053391.1 | NP_445843.1 | 1 | 119 | ||
| GenBank mRNA | AF177430.1 | - | 1 | 119 | ||