siRNA Details
| Gene Symbol: Grb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053403.2 | NP_445855.1 | 2, 3 | 608 | ||
| XM_006247432.2 | XP_006247494.1 | 2, 3 | 2577 | |||
| XM_006247433.2 | XP_006247495.1 | 2, 3 | 460 | |||
| XM_006247434.2 | XP_006247496.1 | 2, 3 | 860 | |||
| XM_008768125.1 | XP_008766347.1 | 2, 3 | 394 | |||
| GenBank mRNA | AF190121.1 | - | 2, 3 | 209 | ||
| BC081757.1 | - | 2, 3 | 608 | 7109771 | ||

Grb7
Genomic Map