siRNA Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109898.1 | NP_001103368.1 | 2 | 316 | ||
| NM_053431.3 | NP_445883.1 | 2 | 316 | |||
| GenBank mRNA | AB044285.1 | - | 2 | 316 | ||
| AF217799.1 | - | 2 | 320 | |||
| AF285078.1 | - | 2 | 338 | |||
| AY623665.1 | - | 2 | 318 | |||

Qsox1
Genomic Map