siRNA Details
| Gene Symbol: Hdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053448.1 | NP_445900.1 | 245 | |||
| GenBank mRNA | AF321131.1 | - | 245 | |||
| BC061988.1 | - | 248 | 5599517 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdac3| Gene Symbol: Hdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053448.1 | NP_445900.1 | 245 | |||
| GenBank mRNA | AF321131.1 | - | 245 | |||
| BC061988.1 | - | 248 | 5599517 | |||